ZRB1956
Anti-Axl Antibody, clone 3D13 ZooMAb® Rabbit Monoclonal

recombinant, expressed in HEK 293 cells
Synonym(s):
Tyrosine-protein kinase receptor UFO;EC:2.7.10.1;AXL oncogene
About This Item
biological source
rabbit
Quality Level
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
3D13, recombinant monoclonal
description
3D13 Clone
product line
ZooMAb® learn more
form
lyophilized
mol wt
calculated mol wt 98.34 kDa
observed mol wt ~140 kDa
purified by
using Protein A
species reactivity
monkey, mouse, human
packaging
antibody small pack of 25 μL
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
sustainability
Greener Alternative Product
technique(s)
affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable
isotype
IgG
epitope sequence
C-terminal
Protein ID accession no.
UniProt accession no.
greener alternative category
shipped in
ambient
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... AXL(558)
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in COS-7 and NIH3T3 cell lysate.
Western Blotting Analysis: A 1:10,000 dilution of this antibody detected Axl (UFO) in COS-7 cell lysate and 1:1,000 dilution detected Axl (UFO) in NIH3T3 cell lysate.
.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Axl in U-251 MG cells.
Flow Cytometry Analysis: 0.1 μg from a representative lot detected Axl (UFO) in one million U251 MG cells.
Affinity Binding Assay: A representative lot of this antibody bound Axl (UFO) peptide with a KD of 3.0 x 10-7 in an affinity binding assay.
Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Axl (UFO) in human skeletal muscle tissue sections.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Reconstitution
Storage and Stability
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service