Skip to Content
MilliporeSigma
All Photos(12)

Key Documents

ZRB1946

Sigma-Aldrich

Anti-Ub-Histone H2A-Lys119 Antibody, clone 8B21 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Histone H2A type 1-A;H2A-clustered histone 1;Histone H2A/r

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

8B21, recombinant monoclonal

description

8B21 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 14.23 kDa
observed mol wt ~20 kDa

purified by

using Protein A

species reactivity

mouse, rat, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
inhibition assay: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

ubiquitination (Lys119)

Gene Information

human ... H2AC1(221613)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 8B21 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Histone H2A ubiqutinylated on lysine 119.

Immunogen

KLH-conjugated linear peptide corresponding to 10 amino acids surrounding ubiquityl lysine 119 from the C-terminal region of human Histone H2A.

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell acid extract.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected Ubiquityl-Histone H2A-Lys119 in HeLa cell acid extract.

Tested Applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected Ubiquityl-Histone H2A-Lys119 in acid exacts of Balb3T3/A31, HL-60, and Rat2 cells.

Peptide Inhibition Assay: Target band detection in HeLa cell acid extract was prevented by preblocking of a representative lot with the immunogen Ubiquityl-Histone H2A-Lys119 peptide.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected Ubiquityl-Histone H2A-Lys119 in human testis tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Ubiquityl-Histone H2A-Lys119 in HeLa cells.

Flow Cytometry Analysis: 1 µg from a representative lot detected Ubiquityl-Histone H2A-Lys119 in one million NIH3T3 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Histone H2A type 1-A (UniProt: Q96QV6; also known as H2A-clustered histone 1, Histone H2A/r) is encoded by the H2AC1 (also known as H2AFR, HIST1H2AA) gene (Gene ID:  221613) in human. Histones are highly conserved basic nuclear proteins that are responsible for the nucleosome structure of chromatin in eukaryotes. They play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which DNA is wrapped in repeating units, called nucleosomes, which limits DNA accessibility to the cellular machineries that require DNA as a template. Histones are modified post-translationally by acetylation, phosphorylation, methylation, and ubiquitination and these modifications regulate DNA transcription, repair, recombination, and replication. Ubiquitylation usually targets the substrate for degradation, although histones H2A and H2B are actually stabilized by a single ubiquitin conjugation. Histone ubiquitination has been correlated with DNA repair and transcription, cellular differentiation, cell cycle regulation, spermatogenesis, protein trafficking, and response to stress. Histone H2A is involved in the structure of chromatin in eukaryotic cells. It features a main globular domain and a long N-terminal tail H2A. Monoubiquitination of lysine 119 (H2AK119Ub) by RING1, TRIM37 and RNF2/RING2 complex, a highly conserved modification, gives a specific tag for epigenetic transcriptional repression and participates in X chromosome inactivation of female mammals. It is involved in the initiation of both imprinted and random X inactivation. Ubiquitinated H2A is reported to be enriched in inactive X chromosome chromatin. H2AK119Ub is also reported to play a role in spermatogenesis and may promote transformation of cells in a number of breast cancers. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Barbour, H., et al. (2020). Nat. Commun. 11; Article 5947; Bhatnagar, S., et al. (2014). Nature. 516(7529); 116-120).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service