Skip to Content
MilliporeSigma
All Photos(9)

Key Documents

ZRB1182

Sigma-Aldrich

Anti-MR-proADM Antibody, clone 2B19 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2B19, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt ~20.42 kDa
observed mol wt ~20 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2B19 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects human ADM. It targets an epitope within the mid-region (MR) of ADM.

Immunogen

KLH-conjugated linear peptide corresponding to 24 amino acids from the propeptide N-terminal half of human ADM.

Application

Quality Control Testing

Evaluated by Western Blotting with recombinant Human ADM Protein.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected recombinant human ADM protein.

Tested Applications

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected MR-proADM in human placenta tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound MR-proADM peptide with a KD of 9.9 x 10-7 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-MR-proADM, clone 2B19 ZooMAb®, Cat. No. ZRB1182, is a recombinant Rabbit monoclonal antibody that specifically targets MR-proADM and is tested for use in Affinity Binding Assay, Immunohistochemistry (Paraffin), and Western Blotting.

Target description

pro-ADM (UniProt: P35318; also known as Mid-regional pro-adrenomedullin) is encoded by the ADM (also known as AM) gene (Gene ID: 133) in human. ADM is synthesized with a signal peptide (aa 1-21) and is cleaved into Adrenomedullin (AM; aa 95-146) and Proadrenomedullin N-20 terminal peptide (PAMP; aa 21-41). AM and PAMP serve as potent hypotensive and vasodilatory agents involved in physiologic control of fluid and electrolyte homeostasis. In the kidney, AM acts as a diuretic and natriuretic, and both AM and PAMP are known to inhibit aldosterone secretion. ADM contains a region known as mid-region ProADM (aa 45-92) that lacks any activity but is a reliable biomarker for sepsis and progression to septic shock. ADM has been found in all epithelial surfaces that separate the internal and external environment, such as integument, respiratory, digestive and genito-urinary tract. It is also found in all bodily secretions and imparts protection against external pathogens. Its highest levels are reported in pheochromocytoma and adrenal medulla. ADM as well as PAMP also display potent antimicrobial action against both Gram-positive and Gram-negative bacteria. MR-proADM, a fragment of 48 amino acids that splits from the proADM and is present in a ratio of 1:1 with ADM, proportionally represents the levels and activity of ADM. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Onal, U., et al. (2018). Healthcare (Basel). 6(3); 110; Valenzuela-Sanchez, F., et al. (2016). Ann. Transl. Med. 4(17); 329).

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service