Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

WH0006935M1

Sigma-Aldrich

Monoclonal Anti-TCF8 antibody produced in mouse

clone 4C4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-AREB6, Anti-BZP, Anti-NIL2A, Anti-PPCD3, Anti-ZEB, Anti-ZEB1, Anti-ZFHEP, Anti-ZFHX1A, Anti-transcription factor 8 (represses interleukin 2 expression)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4C4, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZEB1(6935)

General description

ZEB1 encodes a zinc finger transcription factor that represses T-lymphocyte-specific IL2 gene (MIM 147680) expression by binding to a negative regulatory domain 100 nucleotides 5-prime of the IL2 transcription start site (Williams et al., 1991 [PubMed 1840704]).[supplied by OMIM

Immunogen

ZEB1 (NP_110378, 801 a.a. ~ 900 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
NIAIPTVTAQLPTIVAIADQNSVPCLRALAANKQTILIPQVAYTYSTTVSPAVQEPPLKVIQPNGNQDERQDTSSEGVSNVEDQNDSDSTPPKKKMRKTE

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rosanna Sestito et al.
Communications biology, 3(1), 677-677 (2020-11-15)
Identification of regulatory mechanisms underlying the poor prognosis of ovarian cancer is necessary for diagnostic and therapeutic implications. Here we show that endothelin A receptor (ETAR) and ZEB1 expression is upregulated in mesenchymal ovarian cancer and correlates with poor prognosis.
Toshifumi Murai et al.
Journal of surgical oncology, 109(7), 684-689 (2014-01-24)
Epithelial-to-mesenchymal transition (EMT) is considered to play an important role in cancer invasion and metastasis. The mRNA levels of an epithelial marker (E-cadherin), mesenchymal marker (vimentin), and Zeb-1 were measured in 11 gastric cancer cell lines. Functional analysis was performed
Wei-Ting Huang et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 27(8), 1116-1125 (2014-01-07)
Primary gastric diffuse large B-cell lymphomas may or may not have a concurrent component of mucosa-associated lymphoid tissue lymphoma. Diffuse large B-cell lymphoma/mucosa-associated lymphoid tissue lymphomas are often associated with Helicobacter pylori (H. pylori) infection, suggesting that the large cells
Dandan Yuan et al.
International journal of oncology, 45(6), 2430-2438 (2014-09-10)
Both circulating tumor cells (CTCs) and epithelial-mesenchymal transition (EMT) play an important role in invasion, migration and chemoresistant in tumor development. This study aimed to detect whether EMT occurred in human gastric CTCs and to explore the mechanism of EMT

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service