Key Documents
TMODS3111-0030
Nalgene® friction-fit tube closures
polypropylene, diam. 30 mm, push-fit, case of 20 ea
Synonym(s):
plastic centrifuge tube closures, polypropylene closures
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
polypropylene
packaging
case of 20 ea
manufacturer/tradename
Nalgene DS3111-0030
diam.
30 mm
General description
Easily remove Thermo Scientific Nalgene Friction-Fit Polypropylene Closures for Plastic Centrifuge Tubes from the centrifuge with a convenient pull tab. These autoclavable closures are designed to fit Nalgene round and conical centrifuge tubes. They may not be liquid-tight during high-speed centrifugation.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)