Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP2105

Sigma-Aldrich

SC35 human

recombinant, expressed in insect cells, ≥80% (SDS-PAGE)

Synonym(s):

PR264, SC-35, SC35, SFRS2, SFRS2A, SRp30b

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.26

biological source

human

recombinant

expressed in insect cells

Assay

≥80% (SDS-PAGE)

form

frozen liquid

mol wt

~27 kDa

packaging

pkg of 5 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

650 μg/mL

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SRSF2(6427)

Biochem/physiol Actions

SC35, a member of the SR protein family, is an essential pre-mRNA splicing factor and can function as an essential splicing factor as well as an alternative splicing factor. Phosphorylation of serine residues located within the SR domain directly regulates SC35 activity and compartmentalization of other SR splicing factors. In addition to interacting with RNA and other splicing factors, SC35 has been shown to interact either directly or indirectly with the C-terminal domain (CTD) of the largest subunit of RNA polymerase II, thereby suggesting a potential role of SC35 in coordinating transcription and pre-mRNA splicing.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J F Gui et al.
Proceedings of the National Academy of Sciences of the United States of America, 91(23), 10824-10828 (1994-11-08)
Members of the SR family of pre-mRNA splicing factors are phosphoproteins that share a phosphoepitope specifically recognized by monoclonal antibody (mAb) 104. Recent studies have indicated that phosphorylation may regulate the activity and the intracellular localization of these splicing factors.
James R Boyne et al.
PLoS pathogens, 4(10), e1000194-e1000194 (2008-11-01)
A cellular pre-mRNA undergoes various post-transcriptional processing events, including capping, splicing and polyadenylation prior to nuclear export. Splicing is particularly important for mRNA nuclear export as two distinct multi-protein complexes, known as human TREX (hTREX) and the exon-junction complex (EJC)
A M Zahler et al.
Genes & development, 6(5), 837-847 (1992-05-01)
We demonstrate that four different proteins from calf thymus are able to restore splicing in the same splicing-deficient extract using several different pre-mRNA substrates. These proteins are members of a conserved family of proteins recognized by a monoclonal antibody that
X D Fu et al.
Science (New York, N.Y.), 256(5056), 535-538 (1992-04-24)
The mammalian splicing factor SC35 is required for the first step in the splicing reaction and for spliceosome assembly. The cloning and characterization of a complementary DNA encoding this protein revealed that it is a member of a family of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service