Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP0346

Sigma-Aldrich

EphB2 active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥80% (SDS-PAGE)

Synonym(s):

CAPB, DRT, EK5, EPH receptor B2, ERK

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥80% (SDS-PAGE)

form

aqueous solution

mol wt

52 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

0.5 mg/mL

technique(s)

activity assay: suitable
inhibition assay: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... EPHB2(2048)

General description

Human EPHB2 ( GenBank Accession No. NM_004442), amino acids 569-end with N-terminal His-tag, MW=52 kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Madhu Macrae et al.
Cancer cell, 8(2), 111-118 (2005-08-16)
The EphA2 receptor tyrosine kinase is frequently overexpressed in many cancers, including 40% of breast cancers. Here, we show that EphA2 is a direct transcriptional target of the Ras-Raf-MAPK pathway and that ligand-stimulated EphA2 attenuates the growth factor-induced activation of
Hongshi Xu et al.
American journal of physiology. Renal physiology, 288(4), F855-F866 (2004-11-25)
EphA2, a member of the large family of Eph receptor tyrosine kinases, is highly expressed in epithelial tissue and has been implicated in cell-cell and cell-matrix interactions, as well as cell growth and survival. Expression of EphA2 mRNA and protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service