Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML4125

Ginsenoside Re

new

≥98%

Synonym(s):

(3β,6α,12β)-20-(β-D-Glucopyranosyloxy)-3,12-dihydroxydammar-24-en-6-yl 2-O-(6-deoxy-α-L-mannopyranosyl)-β-D-glucopyranoside, Chikusetsusaponin Ivc, Ginsenoside B2, Notoginsenoside Re, NSC 308877, Panaxoside Re, Re ginsenoside, Sanchinoside Re

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H82O18
CAS Number:
Molecular Weight:
947.15
UNSPSC Code:
12352200

Quality Level

Assay

≥98%

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Active component in ginseng with anti-inflammatory, neuroprotective, antioxidant, cardioprotective, and anticancer properties.

Ginsenoside Re is an active component in ginseng that demonstrates anti-inflammatory effects by modulating pathways and cytokine production, possesses neuroprotective properties against neurodegenerative diseases, exhibits antioxidant activity by neutralizing reactive oxygen species, and shows potential in inhibiting cancer cell growth. Additionally, Ginsenoside-Re has been explored for cardioprotective effects, impacting blood pressure, lipid profiles, and vascular function.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service