Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML4118

Haloperidol decanoate

new

≥98% (HPLC)

Synonym(s):

Decanoic acid 4-(4-chlorophenyl)-1-[4-(4-fluorophenyl)-4-oxobutyl]-4-piperidinyl ester, Haldol decanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H41ClFNO3
CAS Number:
Molecular Weight:
530.11
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Cell-permeable decanoate ester of haloperidol (H1512), a selective and a potent dopamine D2 (DRD2) antagonist, that acts as a long-acting injectable antipsychotic.
Haloperidol decanoate is a cell-permeable decanoate ester of haloperidol (H1512), a selective and a potent dopamine D2 (DRD2) antagonist, that acts as a long-acting injectable antipsychotic. Haloperidol decanoate is reported to be metabolized in the liver and undergo hydrolysis by plasma and/or tissue esterases to form haloperidol and decanoic acid.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service