Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML4110

Echinocystic Acid

new

≥97%

Synonym(s):

(3β,16α)-3,16-Dihydroxyolean-12-en-28-oic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H48O4
CAS Number:
Molecular Weight:
472.70
UNSPSC Code:
12352200

Quality Level

Assay

≥97%

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Pentacyclic triterpene that display numerous activities including anti-tumor, anti-inflammatory, and anti-depression effects; potent inhibitor of Kv7.2/Kv7.3 (KCNQ2-3) channels.
Echinocystic acid is a pentacyclic triterpene extracted from various herbal materials including the fruits of Gleditsia sinensis Lam that display numerous activities including anti-tumor, anti-inflammatory, and anti-depression effects. Echinocystic acid is a potent inhibitor of Kv7.2/Kv7.3 (KCNQ2-3) channels.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service