Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML4090

TH-Z93

new

≥98% (HPLC)

Synonym(s):

(((4-(Hexyloxy)pyridin-2-yl)amino)methylene)bis(phosphonic acid), P,P′-[[[4-(Hexyloxy)-2-pyridinyl]amino]methylene]bis[phosphonic acid]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22N2O7P2
CAS Number:
Molecular Weight:
368.26
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

soluble (H2O + 1 drop 5M NaOH, 2 mg/mL, clear)

storage temp.

2-8°C

Biochem/physiol Actions

Potent mevalonate (MVA) pathway farnesyl diphosphate synthase (FPPS) inhibitor that stimulates immune responses in vivo.


TH-Z93 is a potent farnesyl diphosphate synthase (FPPS) inhibitor (IC50 = 90 nM) that stimulates immune responses by blocking the mevalonate (MVA) pathway-mediated post-translational protein geranylgeranylation. When co-administered with antigen in mice in vivo, TH-Z93 triggers strong adjuvant responses, increasing OVA-specific antibody titres (in mice immunized with 100 μg OVA + 100 μg TH-Z93) and protects against death caused by live influenza virus PR8 challenge (in mice immunized with 5 μg PR8 HA1 subunit containing TH-Z93). TH-Z93 adjuvant efficacy is blocked by GGPP and synergized with HMG-CoA inhibitor simvastatin.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service