Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML4036

Sigma-Aldrich

AN3661

≥98% (HPLC)

Synonym(s):

1,3-Dihydro-1-hydroxy-2,1-benzoxaborole-7-propanoic acid, 3-(1-Hydroxy-1,3-dihydro-2,1-benzoxaborol-7-yl)propanoic acid; 3-(1-Hydroxy-1,3-dihydrobenzo[c][1,2]oxaborol-7-yl)propanoicacid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11BO4
CAS Number:
Molecular Weight:
206.00
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC(CCC1=CC=CC2=C1B(OC2)O)=O

InChI

1S/C10H11BO4/c12-9(13)5-4-7-2-1-3-8-6-15-11(14)10(7)8/h1-3,14H,4-6H2,(H,12,13)

InChI key

CEVOKIPNIZQANN-UHFFFAOYSA-N

Biochem/physiol Actions

Boron containing antimalarial agent that targets Plasmodium falciparum CPSF3 (cleavage and polyadenylation specific factor 3).
AN3661 is a boron containing antimalarial agent that targets Plasmodium falciparum CPSF3 (cleavage and polyadenylation specific factor 3). AN3661potently inhibits Plasmodium falciparum laboratory-adapted strains (mean IC50=32 nM). Also, AN3661 inhibits T. gondii growth in human cells at low micromolar concentrations.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service