Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3937

Sigma-Aldrich

GPX4-Activator-1d4 Hydrochloride

≥98% (HPLC)

Synonym(s):

N-(2-Aminoethyl)-4-(3-cyclopentylthioureido)-benzenesulfonamide hydrochloride, N-(2-Aminoethyl)-4-[[(cyclopentylamino)thioxomethyl]amino]-benzenesulfonamide hydrochloride, PKUMDL-LC-101-D04

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22N4O2S2·HCl
CAS Number:
Molecular Weight:
378.94
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

S=C(NC1=CC=C(C=C1)S(NCCN)(=O)=O)NC2CCCC2.Cl

InChI key

FELDRJSFLBINQS-UHFFFAOYSA-N

Biochem/physiol Actions

Cell penetrant, potent and elective allosteric activator of glutathione peroxidase 4 (GPX4) that potently inhibit ferroptosis.



GPX4-Activator-1d4 HCl is a cell penetrant, potent and elective allosteric activator of glutathione peroxidase 4 (GPX4) that potently inhibit ferroptosis. It significantly increases GPX4 activity to 150% at 20 μM in the cell-free assay and 61 μM in cell extracts. GPX4-Activator-1d4 suppresses ferroptosis and inflammation in HT-1080 fibrosarcoma cells.

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service