SML3858
SAFit2
≥98% (HPLC)
Synonym(s):
(S)-(R)-3-(3,4-Dimethoxyphenyl)-1-(3-(2-morpholinoethoxy)phenyl)propyl-1-((S)-2-cyclohexyl-2-(3,4,5-trimethoxyphenyl)acetyl)piperidine-2-carboxylate, [(1R)-3-(3,4-Dimethoxyphenyl)-1-[3-(2-morpholin-4-ylethoxy)phenyl]propyl] (2S)-1-[(2S)-2-cyclohexyl-2-(3,4,5-trimethoxyphenyl)acetyl]piperidine-2-carboxylate
About This Item
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
-10 to -25°C
SMILES string
O=C([C@@H](C1CCCCC1)C2=CC(OC)=C(C(OC)=C2)OC)N3CCCC[C@H]3C(O[C@@H](C4=CC(OCCN5CCOCC5)=CC=C4)CCC(C=C6)=CC(OC)=C6OC)=O
InChI
1S/C46H62N2O10/c1-51-39-20-18-32(28-40(39)52-2)17-19-38(34-14-11-15-36(29-34)57-27-24-47-22-25-56-26-23-47)58-46(50)37-16-9-10-21-48(37)45(49)43(33-12-7-6-8-13-33)35-30-41(53-3)44(55-5)42(31-35)54-4/h11,14-15,18,20,28-31,33,37-38,43H,6-10,12-13,16-17,19,2
InChI key
ZDBWLRLGUBSLPG-FDHYQTMZSA-N
Biochem/physiol Actions
SAFit2 is a brain-penetrant, potent and selective FK506-binding protein 51 inhibitor (Ki = 6 nM/FKBP51 vs. >50 μM/FKBP52) that enhances neurite growth in murine Neuro 2A (N2a) cultures (1-10 nM) and improves neuroendocrine feedback and stress-coping behavior in mice in vivo (20 mg/kg i.p.).
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service