Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3691

Sigma-Aldrich

Anagrelide analog A6

≥98% (HPLC)

Synonym(s):

6-Chloro-1,5-dihydro-7-(4-methylphenyl)imidazo[2,1-b]quinazolin-2(3H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14ClN3O
CAS Number:
Molecular Weight:
311.77
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Anagrelide analog A6 is a cell penetrant and potent apoptosis inducer that works as a molecular glue bringing together phosphodiesterase 3A (PDE3A) and Schlafen 12 protein (SLFN12) forming heterotetrametric complex. Apparently, anagrelide analog A6 binds to the PDE3A enzymatic pocket that allows recruitment and stabilization of SLFN12, which in turn blocks protein translation, leads to apoptosis. It exhibits potent anticancer activity in cell cultures and in a tumor xenograft model.
Cell penetrant and potent apoptosis inducer that works as a molecular glue bringing together phosphodiesterase 3A (PDE3A) and Schlafen 12 protein (SLFN12)

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT RE 2

Target Organs

Cardiovascular

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service