Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SML3669

Sigma-Aldrich

F-1394

≥98% (HPLC)

Synonym(s):

(1S,2S)-2-(3-neopentyl-3-nonylureido)cyclohexyl 3-((R)-2,2,5,5-tetramethyl-1,3-dioxane-4-carboxamido)propanoate, F 1394, F1394, N-[[(4R)-2,2,5,5-Tetramethyl-1,3-dioxan-4-yl]carbonyl]-β-alanine (1S,2S)-2-[[[(2,2-dimethylpropyl)nonylamino]carbonyl]amino]cyclohexyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H61N3O6
CAS Number:
Molecular Weight:
595.85
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

(Powder or solid or semi-solid or liquid)

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

CCCCCCCCCN(C(N[C@H]1CCCC[C@@H]1OC(CCNC([C@H]2C(C)(COC(C)(O2)C)C)=O)=O)=O)CC(C)(C)C

InChI key

NWLFOBZKYXKBOF-NSVAZKTRSA-N

Biochem/physiol Actions

F1394 is an orally available, potent and selective Acyl-CoA:cholesterol acyltransferase (ACAT) inhibitor that reduces plaque foam cell formation without inflammatory or toxic effects.
Orally available, potent and selective Acyl-CoA:cholesterol acyltransferase (ACAT) inhibitor

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Chiwata et al.
British journal of pharmacology, 133(7), 1005-1012 (2001-08-07)
The acyl-CoA:cholesterol acyltransferase (ACAT) enzyme is thought to be responsible for foam cell formation and the subsequent progression of atherosclerosis. The apolipoprotein E and low density lipoprotein receptor double knockout (apoE/LDLr-DKO) mouse is an animal model that develops severe hyperlipidaemia
Jaume Amengual et al.
Molecular pharmacology, 99(3), 175-183 (2021-01-02)
Acyl-CoA:cholesterol acyltransferase (ACAT) mediates cellular cholesterol esterification. In atherosclerotic plaque macrophages, ACAT promotes cholesteryl ester accumulation, resulting in foam cell formation and atherosclerosis progression. Its complete inactivation in mice, however, showed toxic effects because of an excess of free cholesterol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service