Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3498

Sigma-Aldrich

L6H21

≥98% (HPLC)

Synonym(s):

(2E)-3-(2,3-Dimethoxyphenyl)-1-(4-methoxyphenyl)-2-propen-1-one, (E)-2,3,4′-Trimethoxy-chalcone, (E)-2,3-Dimethoxy-4′-methoxychalcone, L 6H21

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18O4
CAS Number:
Molecular Weight:
298.33
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

L6H21 is an orally bioavailable chalcone that acts as a myeloid differentiation factor 2 (MD2) antagonist and shows a great inhibitory effect on pro-inflammatory cytokines expression levels. L6H21 binds to the hydrophobic pocket of MD2 in a dose-dependent manner with a high affinity (Kd = 33.3 µM) and inhibits the ability of LPS to recognize TLR4/MD2 complex. Suppresses NLRP3 inflammasome activation and may serve as a potential candidate for the treatment of allograft rejection.
Orally bioavailable chalcone that acts as a myeloid differentiation factor 2 (MD2) antagonist

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huaicheng Chen et al.
Experimental & molecular medicine, 53(4), 681-694 (2021-04-21)
Modified LDL-induced inflammation and oxidative stress are involved in the pathogenesis of diabetic retinopathy. Recent studies have also shown that modified LDL activates Toll-like receptor 4 (TLR4) to mediate retinal injury. However, the mechanism by which modified LDL activates TLR4

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service