Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3454

Sigma-Aldrich

Ridaforolimus

≥95% (HPLC)

Synonym(s):

AP23573, Deforolimus, MK-8669

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H84NO14P
CAS Number:
Molecular Weight:
990.21
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O=C(N1CCCC[C@]12[H])C([C@@]3(O)[C@@H](CC[C@]([H])(O3)C[C@H](OC)/C(C)=C/C=C/C=C/[C@@H](C)C[C@H](C([C@@H]([C@H](O)/C(C)=C/[C@H](C(C[C@@]([H])([C@@H](C[C@H]4C[C@@H](OC)[C@@H](CC4)OP(C)(C)=O)C)OC2=O)=O)C)OC)=O)C)C)=O

InChI

1S/C53H84NO14P/c1-32-18-14-13-15-19-33(2)44(63-8)30-40-23-21-38(7)53(61,67-40)50(58)51(59)54-25-17-16-20-41(54)52(60)66-45(35(4)28-39-22-24-43(46(29-39)64-9)68-69(11,12)62)31-42(55)34(3)27-37(6)48(57)49(65-10)47(56)36(5)26-32/h13-15,18-19,27,32,34-36,38-41,43-46,48-49,57,61H,16-17,20-26,28-31H2,1-12H3/b15-13+,18-14+,33-19+,37-27+/t32-,34-,35-,36-,38-,39+,40+,41+,43-,44+,45+,46-,48-,49+,53-/m1/s1

InChI key

BUROJSBIWGDYCN-GAUTUEMISA-N

Biochem/physiol Actions

Antiproliferative mTORC1 inhibitor
Ridaforolimus is an antiproliferative mTOR inhibitor that forms a complex with the FK506 binding protein-12 (FKBP-12), which binds to and inhibits mTOR resulting in cell cycle arrest and inhibition of tumor cell growth and proliferation. Like rapamycin, ridaforolimus affects only mTORC1, not mTORC2.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service