Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3184

Sigma-Aldrich

Santacruzamate A

≥98% (HPLC)

Synonym(s):

CAY 10683, CAY-10683, CAY10683, N-[4-Oxo-4-[(2-phenylethyl)amino]butyl]carbamic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22N2O3
CAS Number:
Molecular Weight:
278.35
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(CCCNC(OCC)=O)NCCC1=CC=CC=C1

InChI

1S/C15H22N2O3/c1-2-20-15(19)17-11-6-9-14(18)16-12-10-13-7-4-3-5-8-13/h3-5,7-8H,2,6,9-12H2,1H3,(H,16,18)(H,17,19)

InChI key

HTOYBIILVCHURC-UHFFFAOYSA-N

Biochem/physiol Actions

Santacruzamate A (CAY10683) is a cell-permeable and bioavailable carbamate compound that potently inhibits HDAC2 (IC50 = 119 nM) selectively over HDAC4 and HDAC6 (IC50 = > 1 μM and 433.5 nM, respectively). CAY10683 readily enhances cellular acetylated HDAC2 levels upon LPS-stimulation and only modestly inhibits HuT-78 cutaneous T-cell lymphoma and HCT-116 colon carcinoma cells growth (GI50 = 1.4 and 29.4 uM, respectively). It alleviates acute liver failure (ALF) in rats (i.p.) and rescues cognitive deficits in APPswe/PS1dE9 mice (p.o.).
cell-permeable and bioavailable carbamate compound that potently inhibits HDAC2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service