Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2501

Sigma-Aldrich

MF-094

≥98% (HPLC)

Synonym(s):

(S)-N-(1-((5-(N-(tert-butyl)sulfamoyl)naphthalen-1-yl)amino)-1-oxo-3-phenylpropan-2-yl)cyclohexanecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H37N3O4S
CAS Number:
Molecular Weight:
535.70
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(NC(C)(C)C)c1c2c(c(ccc2)NC(=O)[C@@H](NC(=O)C4CCCCC4)Cc3ccccc3)ccc1

InChI key

DQXORJHBZHLLTE-SANMLTNESA-N

Biochem/physiol Actions

Inhibitor of ubiquitin specific protease 30 (USP30)
MF-094 is a potent inhibitor of ubiquitin specific protease 30 (USP30), a de-ubiquitinating enzymes (DUB) that opposes the action of parkin-mediated ubiquitination in clearing damaged mitochondria. MF-094 is selective for USP30 with an IC50 value of 120 nM, compared to >10 μM for USPs 1, 8, and 9.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arthur F Kluge et al.
Bioorganic & medicinal chemistry letters, 28(15), 2655-2659 (2018-06-25)
Mitophagy is one of the processes that cells use to maintain overall health. An E3 ligase, parkin, ubiquitinates mitochondrial proteins prior to their degradation by autophagasomes. USP30 is an enzyme that de-ubiquitinates mitochondrial proteins; therefore, inhibiting this enzyme could foster
Hang Zhu et al.
Redox biology, 38, 101777-101777 (2020-11-10)
The death of cardiomyocytes either through apoptosis or necroptosis is the pathological feature of cardiac ischemia-reperfusion (I/R) injury. Phosphoglycerate mutase 5 (PGAM5), a mitochondrially-localized serine/threonine-protein phosphatase, functions as a novel inducer of necroptosis. However, intense debate exists regarding the effect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service