SML2113
FR194921
≥98% (HPLC)
Synonym(s):
2-(1-Methyl-4-piperidinyl)-6-[2-phenylpyrazolo-(1,5-a)pyridin-3-yl]-3(2H)-pyridazinone
About This Item
Recommended Products
Assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear (warmed)
storage temp.
2-8°C
SMILES string
CN(CC1)CCC1N2N=C(C=CC2=O)C3=C4N(N=C3C5=CC=CC=C5)C=CC=C4
InChI
1S/C23H23N5O/c1-26-15-12-18(13-16-26)28-21(29)11-10-19(24-28)22-20-9-5-6-14-27(20)25-23(22)17-7-3-2-4-8-17/h2-11,14,18H,12-13,15-16H2,1H3
InChI key
YHDRUTMZCJZJAL-UHFFFAOYSA-N
Biochem/physiol Actions
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service