Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1498

Sigma-Aldrich

Cinepazide maleate

≥98% (HPLC)

Synonym(s):

(E)-1-[4-(2-Oxo-2-pyrrolidin-1-ylethyl)piperazin-1-yl]-3-(3,4,5-trimethoxyphenyl)prop-2-en-1-one maleate, 1-(Pyrrolidinyl-1-carbonyl)-methyl-4-(3,4,5-trimethoxycinnamoyl)-piperazine-maleate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H31N3O5 · C4H4O4
CAS Number:
Molecular Weight:
533.57
MDL number:
UNSPSC Code:
12352125
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(O)/C=C\C(O)=O.O=C(N1CCN(CC(N2CCCC2)=O)CC1)/C=C/C3=CC(OC)=C(OC)C(OC)=C3

InChI

1S/C22H31N3O5.C4H4O4/c1-28-18-14-17(15-19(29-2)22(18)30-3)6-7-20(26)25-12-10-23(11-13-25)16-21(27)24-8-4-5-9-24;5-3(6)1-2-4(7)8/h6-7,14-15H,4-5,8-13,16H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b7-6+;2-1-

InChI key

XSTJTOKYCAJVMJ-GVTSEVKNSA-N

General description

Cinepazide maleate can be synthesized from ethyl chlorocarbonate activation and (E) 3,4,5-trimethoxycinnamylic acid.

Biochem/physiol Actions

Cinepazide maleate is a vasodilator used clinically in China used for the treatment of cardiovascular and cerebrovascular diseases, and peripheral vascular diseases.
Cinepazide maleate is a vasodilator used clinically in China used in China for the treatment of cardiovascular and cerebrovascular diseases, and peripheral vascular diseases. Cinepazide maleate is thought to act as a potentiator of adenosine A2 receptors and has also been characterized as a calcium channel blocker. It was withdrawn for agranulocytosis in several countries.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An Improved Synthesis of Cinepazide Maleate.
LIU, Hua-xiang, et al.
Fine Chemicals / ????, 10, 012-012 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service