Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0229

Sigma-Aldrich

CIQ

≥98% (HPLC)

Synonym(s):

(3-Chlorophenyl)[3,4-dihydro-6,7-dimethoxy-1-[(4-methoxyphenoxy)methyl]-2(1H)-isoquinolinyl]-methanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H26ClNO5
CAS Number:
Molecular Weight:
467.94
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: ≥10 mg/mL at warmed to 60 °C

storage temp.

2-8°C

SMILES string

Clc1cc(ccc1)C(=O)N2CCc3c(cc(c(c3)OC)OC)C2COc4ccc(cc4)OC

InChI

1S/C26H26ClNO5/c1-30-20-7-9-21(10-8-20)33-16-23-22-15-25(32-3)24(31-2)14-17(22)11-12-28(23)26(29)18-5-4-6-19(27)13-18/h4-10,13-15,23H,11-12,16H2,1-3H3

InChI key

VYMILMYEENZHAR-UHFFFAOYSA-N

Biochem/physiol Actions

CIQ is a selective NR2C/NR2D-containing NMDA potentiator
CIQ is positive allosteric modulator of NR2C/NR2D subunit containing NMDA receptors. CIQ increases channel opening frequency of recombinant NR2Cor NR2D containing receptors by two-fold (EC50 = 2.7 and 2.8 μM, respectively), with no effect on NR2A or NR2B subtypes. The compound does not alter the EC50 values for glutamate or glycine on channel opening.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Glutamate Receptors (Ion Channel Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offers many products related to ion channel family glutamate receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service