Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCP0131

Sigma-Aldrich

ECE-1 Substrate

≥95% (HPLC), lyophilized

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C66H81N15O19
Molecular Weight:
1388.44
UNSPSC Code:
12352204
NACRES:
NA.32

Product Name

ECE-1 Substrate, ≥95% (HPLC)

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥50%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

MCA-Arg-Pro-Pro-Gly-Phe-Ser-Ala-Phe-DNP-Lys

Application

MCA-RPPGFSAF-DNP-K (Mca-RPPGFSAFK(Dnp)) is a fluorogenic substrate used to assay Angiotensin converting enzyme (ACE-1), insulin-degrading enzyme and Endothelin converting enzyme 1 (ECE-1) activities.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeff C Joyner et al.
Journal of the American Chemical Society, 134(7), 3396-3410 (2011-12-28)
A series of compounds that target reactive transition-metal chelates to somatic angiotensin converting enzyme (sACE-1) have been synthesized. Half-maximal inhibitory concentrations (IC(50)) and rate constants for both inactivation and cleavage of full-length sACE-1 have been determined and evaluated in terms
James Scott Miners et al.
Journal of neuroscience methods, 169(1), 177-181 (2008-01-29)
Internally quenched fluorogenic substrates are commonly used for measuring enzyme activity in biological samples and allow high sensitivity and continuous real-time measurement that is well suited for high throughput analysis. We describe the development and optimisation of an immunocapture-based assay
Burcu Hasdemir et al.
Molecular endocrinology (Baltimore, Md.), 26(4), 681-695 (2012-02-11)
CRF receptor 1 (CRF(1)), a key neuroendocrine mediator of the stress response, has two known agonists corticotropin-releasing factor (CRF) and urocortin 1 (Ucn1). Here we report that endothelin-converting enzyme-1 (ECE-1) differentially degrades CRF and Ucn1; ECE-1 cleaves Ucn1, but not
Juan-Carlos Pelayo et al.
The Journal of physiology, 589(Pt 21), 5213-5230 (2011-09-01)
Neuropeptide signalling at the plasma membrane is terminated by neuropeptide degradation by cell-surface peptidases, and by β-arrestin-dependent receptor desensitization and endocytosis. However, receptors continue to signal from endosomes by β-arrestin-dependent processes, and endosomal sorting mediates recycling and resensitization of plasma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service