Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SBR00098

Sigma-Aldrich

Tazemetostat

≥98% (HPLC), powder

Synonym(s):

EPZ6438

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H44N4O4
CAS Number:
Molecular Weight:
572.74
UNSPSC Code:
51101500

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

Mode of action

protein synthesis | inhibits

storage temp.

2-30°C

SMILES string

CCN(C1CCOCC1)C2=CC(=CC(=C2C)C(=O)NCC3=C(C=C(NC3=O)C)C)C4=CC=C(C=C4)CN5CCOCC5

InChI

InChI=1S/C34H44N4O4/c1-5-38(29-10-14-41-15-11-29)32-20-28(27-8-6-26(7-9-27)22-37-12-16-42-17-13-37)19-30(25(32)4)33(39)35-21-31-23(2)18-24(3)36-34(31)40/h6-9,18-20,29H,5,10-17,21-22H2,1-4H3,(H,35,39)(H,36,40)

InChI key

NSQSAUGJQHDYNO-UHFFFAOYSA-N

General description

Tazemetostat is a small molecule inhibitor of the EZH2 protein, which is a histone-lysine N-methyltransferase enzyme that plays a key role in epigenetic regulation of gene expression.

Biochem/physiol Actions

Tazemetostat inhibits the EZH2 protein. EZH2 is a histone-lysine N-methyltransferase enzyme that plays a key role in epigenetic regulation of gene expression in cells. As an epigenetic modifier, EZH2 protein controls the transcriptional repression of various genes involved in cell cycle regulation, DNA repair, and other cellular processes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service