Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB5702747

Sigma-Aldrich

Anti-KAT13A / SRC1 Antibody, clone 3S4C6, Rabbit Monoclonal

Synonym(s):

F-SRC-1, KAT13A, RIP160, SRC1, bHLHe42, bHLHe74

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

3S4C6, monoclonal

form

liquid

mol wt

180 kDa

species reactivity

human

concentration

1.55 mg/mL

technique(s)

immunoprecipitation (IP): 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

VARSSTLPPSNSNMVSTRINRQQSSDLHSSSHSNSSNSQGSFGCSPGSQIVANVALNQGQASSQSSNPSLNLNNSPMEGTGISLAQFMSPRRQVTSGLATRPRMPNNSFPPNISTLSSPVGMTSSACNNNNRSYSNIPVTSLQGMNEGPNNSVGFSASSPVLRQMSSQNSPSRLNIQPAKAESKDNKEIASILNEMIQSDNSSSDGKPLDSGLLHNNDRLSDGDSKYSQTSHKLVQLLTTTAEQQLRHADIDTSCKDVLSCTGTSNSASANSSGGSCPSSHSSLTERHKILHRLLQEGSPSDITTLSVEPDKKDSASTSVSVTGQVQGNSSIKLELDASKKKESKDHQLLR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NCOA1(8648)

General description

The protein encoded by this gene acts as a transcriptional coactivator for steroid and nuclear hormone receptors. It is a member of the p160/steroid receptor coactivator (SRC) family and like other family members has histone acetyltransferase activity and contains a nuclear localization signal, as well as bHLH and PAS domains. The product of this gene binds nuclear receptors directly and stimulates the transcriptional activities in a hormone-dependent fashion. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq, Jul 2008]

Immunogen

PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.
Recombinant fusion protein containing a sequence corresponding to amino acids 400-750 of human KAT13A / SRC1 (Q15788).

Application

WB, IP

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service