Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB5500135

Sigma-Aldrich

Anti-KU80 antibody, Rabbit monoclonal

clone SP130, recombinant, expressed in proprietary host, affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in proprietary host

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

SP130, monoclonal

species reactivity

human (tested)

species reactivity (predicted by homology)

bovine, rat, mouse

technique(s)

immunoblotting: 1:25
immunohistochemistry: 1:100

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... XRCC5(7520)

General description

X-ray repair cross-complementing protein 5 (XRCC5), alternative name (Ku80), is single stranded DNA-dependent ATP-dependent helicase. It involves in chromosome translocation. Ku80 along with Ku70 forms a heterodimeric protein originally identified as autoantigens associated with several autoimmune diseases including scleroderma, polymyositis and systemic lupus erythematosus. Ku is widely expressed nuclear protein that binds to and stabilizes the ends of DNA at telomeres or double-stranded DNA breaks. The inhibition of Ku80 can damage the DNA repair pathway of tumor cells and potentially increase the killing of cancer cells, and reduce drug resistance.

Immunogen

Synthetic peptide corresponding to an internal region within Human Ku80 aa 700 to the C-terminus.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1 ml rabbit monoclonal antibody purified by protein A/G in PBS/1% BSA buffer pH 7.6 with less than 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service