Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4702041

Monoclonal Anti-HLA-DR APC-Cyanine7 produced in mouse

new
Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

conjugate

APC-Cy7 conjugate

Quality Level

antibody product type

primary antibodies

clone

L243, monoclonal

form

buffered aqueous solution

species reactivity

( Non-human primates), ( Human), dog

technique(s)

flow cytometry: suitable

isotype

IgG2aκ

storage temp.

2-8°C

target post-translational modification

unmodified

General description

HLA-DR, a member of MHC class II glycoproteins, that bind intracellularly processed peptides and present them to the Th cells, is composed of 36 kDa alpha chain and 27 kDa beta chain, both anchored in the plasma membrane. Together with other MHC II molecules HLA-DR plays a central role in the immune system.

Specificity

The mouse monoclonal antibody L243 recognizes specifically an extracellular epitope on HLA-DR molecules, both peptide-loaded and empty.

Immunogen

Human B lymphocytes

Physical form

Stabilizing phosphate buffered saline (PBS), pH 7.4, 15 mM sodium azide

Preparation Note

Purified antibody is conjugated with activated tandem dye of allophycocyanin-cyanine 7 (APC-Cy7) under optimum conditions and unconjugated antibody and free fluorochrome are removed by size-exclusion chromatography.

Storage and Stability

Store at 2-8°C. Protect from prolonged exposure to light. Do not freeze.

Other Notes

The antibody solution should be gently mixed before use.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service