Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4700220

Sigma-Aldrich

Monoclonal Anti-CD69-APC antibody produced in mouse

clone FN50, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

Allophycocyanin conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

FN50, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CD69(969)

General description

Cluster of differentiation 69 (CD69) is a type II integral membrane protein, encoded by the gene mapped to human chromosome 12p13.31. CD69 is a member of the C-type lectin family of surface receptors. The encoded protein is expressed in all bone marrow-derived cells, with the exception of erythrocytes.
The antibody FN50 recognizes CD69, an lymphocyte early activation marker.

Immunogen

anti-mju-stimulated human B lymphocyts

Application

The reagent is designed for Flow Cytometry analysis of human blood cells using 10 μL reagent / 100 μL of whole blood or 1e6 cells in a suspension. The content of a vial (1 mL) is sufficient for 100 tests.

Biochem/physiol Actions

Cluster of differentiation 69 (CD69) exerts a pro-inflammatory effect by activating leukocytes. In humans, CD69 negatively regulates immune reactivity. Knockout/ deficiency of the gene increases the risk of susceptibility to autoimmune and chronic inflammatory diseases. The encoded protein might act as a prognostic marker for bendamustine response in chronic lymphocytic leukemia (CLL) patients. CD69 facilitates natural killer (NK) cell function. In addition it also helps in activation and differentiation of a wide variety of hematopoietic cells.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline containing 15 mM sodium azide and 0.2% high-grade protease free BSA as a stabilizing agent.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Is CD69 an effective brake to control inflammatory diseases?
Gonzalez-Amaro R
Trends in Molecular Medicine, 19, 625-632 (2013)
CD69 expression potentially predicts response to bendamustine and its modulation by ibrutinib or idelalisib enhances cytotoxic effect in chronic lymphocytic leukemia.
Montraveta A
Oncotarget, 7, 5507-5520 (2016)
The activation antigen CD69
Ziegler SF
Stem Cells, 12, 456-465 (1994)
Genome-wide association study and meta-analysis finds over 40 loci affect risk of type 1 diabetes
Barrett JC
Nature Genetics, 41, 703-707 (2009)
Costin Tomescu et al.
Journal of immunology (Baltimore, Md. : 1950), 179(4), 2097-2104 (2007-08-07)
In vivo, several mechanisms have been postulated to protect HIV-1-infected cells from NK surveillance. In vitro, previous research indicates HIV-1-infected autologous CD4(+) primary T cells are resistant to NK lysis. We hypothesized that NK lysis of HIV-1-infected target cells would

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service