Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4300565

Sigma-Aldrich

Anti-SMAD3 (Ab-425) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-DKFZp586N0721 antibody produced in rabbit, Anti-DKFZp686J10186 antibody produced in rabbit, Anti-HSPC193 antibody produced in rabbit, Anti-HsT17436 antibody produced in rabbit, Anti-SMAD family member 3 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~52 kDa

species reactivity

human, mouse, rat

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
indirect immunofluorescence: 1:100-1:200

isotype

IgG

immunogen sequence

(C-S-S-V-S)

NCBI accession no.

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SMAD3(4088)

Immunogen

Peptide sequence around aa. 423-427 (C-S-S-V-S), according to the protein SMAD3.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Smad3 encoded by this gene belongs to the SMAD, a family of proteins similar to the gene products of the Drosophila gene 'mothers against decapentaplegic' (Mad) and the C. elegans gene Sma. SMAD proteins are signal transducers and transcriptional modulators that mediate multiple signaling pathways. This protein functions as a transcriptional modulator activated by transforming growth factor-beta and is thought to play a role in the regulation of carcinogenesis.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soo Youn Cho et al.
Medical oncology (Northwood, London, England), 31(11), 236-236 (2014-10-01)
Smad3 functions as an integrator of diverse signaling, including transforming growth factor β signaling and the function of Smad3 is complexly regulated by differential phosphorylation at various sites of Smad3. Despite the importance of Smad3 and its various phosphoisoforms, their
Nicolás Tobar et al.
BMC cancer, 14, 640-640 (2014-09-02)
Hard consistency, developed under the influence of tumor cell factors, is a characteristic feature of a breast tumor. Activation of resident fibroblasts leading to a myofibroblast phenotype is the principal feature that orchestrates this fibrotic process. The aim of this
Jinyi Han et al.
Experimental biology and medicine (Maywood, N.J.), 239(3), 272-283 (2014-02-07)
All-trans retinoic acid (ATRA) has been used for the treatment of acute promyelocytic leukemia. It remains unclear, however, whether ATRA affects cyclooxygenase-2 (COX-2; an enzyme involved in prostaglandin production), PGE2, and thromboxane A2 (TXA2) (metabolic products of COX-2) by a
Yunchuan Li et al.
Chinese medical journal, 127(19), 3449-3453 (2014-10-02)
Aberrant epithelial repair has been observed in chronic rhinosinusitis (CRS) patients; however, the mechanism of epithelial cell repair regulation is unclear. Epidermal growth factor (EGF) plays an important role in regulating epithelial cell repair in lower airway and may be
David Adler et al.
Endocrine-related cancer, 21(4), 663-675 (2014-06-19)
In a recent effort to unravel the molecular basis of prostate cancer (PCa), Barbieri and colleagues using whole-exome sequencing identified a novel recurrently mutated gene, MED12, in 5.4% of primary PCa. MED12, encoding a subunit of the Mediator complex, is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service