Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4300158

Sigma-Aldrich

Anti-phospho-HSP90AB1 (pSer254) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-D6S182 antibody produced in rabbit, Anti-FLJ26984 antibody produced in rabbit, Anti-HSP90-BETA antibody produced in rabbit, Anti-HSP90B antibody produced in rabbit, Anti-heat shock protein 90kDa alpha (cytosolic), class B member 1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~92 kDa

species reactivity

mouse, human

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(V-G-SP-D-E)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer254)

Gene Information

human ... HSP90AB1(3326)

Immunogen

Peptide sequence around phosphorylation site of serine 254 (V-G-S(p)-D-E), according to the protein HSP90AB1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Molecular chaperone. Has ATPase activity.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service