SAB4200848
Anti-FAM134B (lumenal) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution
Synonym(s):
JK1, RETREG1, reticulophagy regulator 1
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
Recommended Products
antibody form
affinity isolated antibody
Quality Level
form
liquid
species reactivity
human
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
concentration
~1 mg/mL
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Related Categories
General description
Five endoplasmic reticulum (ER)-resident proteins have been shown to function as receptors for selective ER-phagy: FAM134B, SEC62, RTN3, CCPG1 and ATL3.1-2 These integral membrane proteins appear to partition into different sub-domains of the ER. All of them harbor a sequential peptide motif within the cytoplasmic region, enabling binding to LC3/GABARAP proteins associated with phagophore membrane.1 FAM134B (also known as RETREG1 or JK-1) is the first ER-phagy receptor to be identified.2
Specificity
Anti-FAM134B (lumenal) antibody specifically recognizes an epitope on human FAM134B ER luminal domain.
Application
The antibody may be used in various immunochemical techniques including Immunoblotting (~55 kDa) and Immunofluorescence. Detection of the FAM134B band by Immunoblotting is specifically inhibited by the immunogen.
Biochem/physiol Actions
FAM134B was discovered to be involved in the pathogenesis of esophageal squamous cell carcinoma (ESCC).3-4 Loss of function mutations in FAM134B result in severe sensory and autonomic neuropathy (HSANII).5 In addition, FAM134B has been classified as an intra-membrane ER-resident protein that is characterized by the presence of a reticulon homology domain (RHD) and that is mainly located at the edges of the ER sheets.1,6 In research, FAM134B is implicated in the suppression of viral replication during Ebola, Dengue, Zika and West Nile viral infections.7-8 Non-structural viral proteases such as NS2S3 specifically cleave FAM134B, thereby subverting ER-phagy.7 Specific mutations in FAM134B and expression profiles are strongly correlated with colorectal cancers.9 Increased expression of FAM134B is also implicated with susceptibility to vascular dementia and allergic rhinitis.9-10
Physical form
Supplied as a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative.
Storage and Stability
For continuous use, store at 2-8°C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.
Disclaimer
Unless otherwise stated in our catalog our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
J C Tang et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 7(6), 1539-1545 (2001-06-19)
In this study, we screened 19 esophageal squamous cell carcinomas (ESCCs) for the detection of genetic alterations using inter-simple sequence repeat PCR, a DNA fingerprinting approach. Three simple repetitive unanchored primers representing tri- and tetranucleotide repeats [(GTG)(5), (GACA)(4), and (GATA)(4)]
Nicholas J Lennemann et al.
Autophagy, 13(2), 322-332 (2017-01-20)
The endoplasmic reticulum (ER) is exploited by several diverse viruses during their infectious life cycles. Flaviviruses, including dengue virus (DENV) and Zika virus (ZIKV), utilize the ER as a source of membranes to establish their replication organelles and to facilitate
Ingo Kurth et al.
Nature genetics, 41(11), 1179-1181 (2009-10-20)
Hereditary sensory and autonomic neuropathy type II (HSAN II) leads to severe mutilations because of impaired nociception and autonomic dysfunction. Here we show that loss-of-function mutations in FAM134B, encoding a newly identified cis-Golgi protein, cause HSAN II. Fam134b knockdown results
Paolo Grumati et al.
Journal of cell science, 131(17) (2018-09-05)
Selective autophagy represents the major quality control mechanism that ensures proper turnover of exhausted or harmful organelles, among them the endoplasmic reticulum (ER), which is fragmented and delivered to the lysosome for degradation via a specific type of autophagy called
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service