Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2900429

Sigma-Aldrich

Anti-GPR35 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

G protein-coupled receptor 35, G-protein coupled receptor 35, GPR35, KYNA receptor, Kynurenic acid receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1.25-2.5 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... GPR35(2859)

General description

ARTS1/Adipocyte-derived leucine aminopeptidase (A-LAP) is a member of the M1 family of zinc metallopeptidases. A-LAP is upregulatable by gamma interferon and trims N-terminal lysine, leucine, tyrosine and asparagines residues when they are not followed by proline in the ER to yield the optimal octamer, primarily and nonamer peptides for antigen presentation by major histocompatibility complex (MHC) molecules. It may also play a role in the inactivation of peptide hormones and may be involved in the regulation of blood pressure through the inactivation of angiotensin II and/or the generation of bradykinin in the kidney.

Immunogen

Synthetic 18 amino acid peptide from 3rd extracellular domain of human GPR35. Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon (100%); Monkey (83%).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate-buffered saline containing less than 0.1% sodium azide.
Supplied at 1 mg/ml

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service