Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2500833

Sigma-Aldrich

Anti-PSME3 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Ki, Anti-Macropain activator subunit 3, Anti-PA28-γ, Anti-PA28G

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human, mouse, canine

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PSME3(10197)

Immunogen

Peptide with sequence C-KNIEKIKRPRSSN from the C Terminus of the protein sequence according to NP_005780.2; NP_789839.1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Li et al.
Journal of molecular biology, 299(3), 641-654 (2000-06-03)
The REG homologs, alpha, beta and gamma, activate mammalian proteasomes in distinct ways. REGalpha and REGbeta activate the trypsin-like, chymotrypsin-like and peptidylglutamyl-preferring active sites, whereas REGgamma only activates the proteasome's trypsin-like subunit. The three REG homologs differ in carboxyl-terminal sequences
S Wilk et al.
Archives of biochemistry and biophysics, 383(2), 265-271 (2001-02-24)
PA28 or 11S REG is a proteasome activator composed of homologous alpha- and beta-subunits and predominantly found in the cytosol. A homologous protein originally known as the Ki antigen but now called PA28gamma or REGgamma is predominantly localized in the
Martina Magni et al.
Nucleic acids research, 42(21), 13150-13160 (2014-11-02)
Human DBC1 (Deleted in Breast Cancer 1; KIAA1967; CCAR2) is a protein implicated in the regulation of apoptosis, transcription and histone modifications. Upon DNA damage, DBC1 is phosphorylated by ATM/ATR on Thr454 and this modification increases its inhibitory interaction with
Xiaolin Gao et al.
Archives of biochemistry and biophysics, 425(2), 158-164 (2004-04-28)
The proteasome activation properties of recombinant REG gamma molecules depend on purification procedures. Prior to ammonium sulfate precipitation recombinant REG gamma activates the trypsin-like catalytic subunit of the proteasome; afterwards it activates all three catalytic subunits. The expanded activation specificity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service