Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SAB1412386

Sigma-Aldrich

Anti-MAPK3/ERK1 Antibody

mouse monoclonal, 1E2

Synonym(s):

ERK1, HS44KDAP, HUMKER1A, MAPK3, MGC20180

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

ANTI-MAPK3 antibody produced in mouse, clone 1E2, purified immunoglobulin, buffered aqueous solution

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1E2, monoclonal

form

buffered aqueous solution

mol wt

antigen 36.74 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAPK3(5595)

General description

The protein encoded by this gene is a member of the MAP kinase family. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act in a signaling cascade that regulates various cellular processes such as proliferation, differentiation, and cell cycle progression in response to a variety of extracellular signals. This kinase is activated by upstream kinases, resulting in its translocation to the nucleus where it phosphorylates nuclear targets. Alternatively spliced transcript variants encoding different protein isoforms have been described. (provided by RefSeq)

Immunogen

MAPK3 (AAH13992, 279 a.a. ~ 379 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
NYLQSLPSKTKVAWAKLFPKSDSKALDLLDRMLTFNPNKRITVEEALAHPYLEQYYDPTDEPVAEEPFTFAMELDDLPKERLKELIFQETARFQPGVLEAP

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haizhong Feng et al.
The Journal of clinical investigation, 124(9), 3741-3756 (2014-07-26)
Aberrant activation of EGFR in human cancers promotes tumorigenesis through stimulation of AKT signaling. Here, we determined that the discoidina neuropilin-like membrane protein DCBLD2 is upregulated in clinical specimens of glioblastomas and head and neck cancers (HNCs) and is required
Eszter Banki et al.
Journal of molecular neuroscience : MN, 54(3), 300-309 (2014-02-19)
Diabetic nephropathy is the leading cause of end-stage renal failure and accounts for 30-40 % of patients entering renal transplant programmes. The nephroprotective effects of the neuropeptide pituitary adenylate cyclase-activating polypeptide (PACAP38) against diabetes have been shown previously, but the molecular
Ning Chai et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 39(8), 1933-1945 (2014-02-21)
Fear extinction has been extensively studied, but little is known about the molecular processes that underlie the persistence of extinction long-term memory (LTM). We found that microinfusion of norepinephrine (NE) into the CA1 area of the dorsal hippocampus during the
Jessica Tilghman et al.
Cancer research, 74(11), 3168-3179 (2014-04-09)
Glioblastoma (GBM) stem cells (GSC) are a subpopulation of tumor cells that display stem-like characteristics (stemness) and play unique roles in tumor propagation, therapeutic resistance, and tumor recurrence. Therapeutic targets in GSCs are a focus of increasing interest to improve
Yichao Fan et al.
Experimental eye research, 127, 104-116 (2014-06-10)
The breakdown of the inner endothelial blood-retinal barrier (BRB) and subsequent retinal vascular leakage are the main causes of vision loss due to diabetic retinopathy (DR). Exendin-4 (E4) is a long-acting agonist of the glucagon-like peptide 1 hormone receptor (GLP-1R)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service