Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1411363

Sigma-Aldrich

Anti-NR2C2 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

TAK1, TR2R1, TR4, hTAK1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 57.5 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NR2C2(7182)

General description

Members of the nuclear hormone receptor family, such as NR2C2, act as ligand-activated transcription factors. The proteins have an N-terminal transactivation domain, a central DNA-binding domain with 2 zinc fingers, and a ligand-binding domain at the C terminus. The activated receptor/ligand complex is translocated to the nucleus where it binds to hormone response elements of target genes (Yoshikawa et al., 1996 [PubMed 8661150]).[supplied by OMIM

Immunogen

NR2C2 (AAH51670.1, 1 a.a. ~ 530 a.a) full-length human protein.

Sequence
MTSPSPRIQIISTDSAVASPQRIQGSEPASGPLSVFTSLNKEKIVTDQQTGQKIQIVTAVDASGSPKQQFTLTSPDGAGTGKVILASPETSSAKQLIFTTSDNLVPGRIQIVTDSASVERLLGKTDVQRPQVVEYCVVCGDKASGRHYGAVSCEGCKGFFKRSVRKNLTYSCRSNQDCIINKHHRNRCQFCRLKKCLEMGMKMESVQSKRKPFDVQREKPSNCAASTEKIYIRKDLRSPLIATPTFVADKDGARQTGLLDPGMLVNIQQPLIREDGTVLLATDSKAETSQGALGTLANVVTSLANLSESLNNGDTSEIQPEDQSASEITRAFDTLAKALNTTDSSSSPSLADGIDTSGGGSIHVISRDQSTPIIEVEGPLLSDTHVTFKLTMPSPMPEYLNVHYICESASRLLFLSMHWARSIPAFQALGQDCNTSLVRACWNELFTLGLAQCAQVMSLSTILAAIVNHLQNSIQEDKLSGDRIKQVMEHIWKLQEFCNSMAKLDIDGYEYAYLKAIVLFSPGKICGGDA

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shin-Jen Lin et al.
Carcinogenesis, 35(6), 1399-1406 (2014-03-04)
Testicular nuclear receptor 4 (TR4), a member of the nuclear receptor superfamily, plays important roles in metabolism, fertility and aging. The linkage of TR4 functions in cancer progression, however, remains unclear. Using three different mouse models, we found TR4 could

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service