Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1411275

Sigma-Aldrich

Anti-ST3GAL2 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

Gal-NAc6S, SIAT4B, ST3GALII, ST3GalA.2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 40.2 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ST3GAL2(6483)

General description

The protein encoded by this gene is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. The encoded protein is normally found in the Golgi but can be proteolytically processed to a soluble form. This protein, which is a member of glycosyltransferase family 29, can use the same acceptor substrates as does sialyltransferase 4A. (provided by RefSeq)

Immunogen

ST3GAL2 (NP_008858.1, 1 a.a. ~ 350 a.a) full-length human protein.

Sequence
MKCSLRVWFLSVAFLLVFIMSLLFTYSHHSMATLPYLDSGALDGTHRVKLVPGYAGLQRLSKERLSGKSCACRRCMGDAGASDWFDSHFDGNISPVWTRENMDLPPDVQRWWMMLQPQFKSHNTNEVLEKLFQIVPGENPYRFRDPHQCRRCAVVGNSGNLRGSGYGQDVDGHNFIMRMNQAPTVGFEQDVGSRTTHHFMYPESAKNLPANVSFVLVPFKVLDLLWIASALSTGQIRFTYAPVKSFLRVDKEKVQIYNPAFFKYIHDRWTEHHGRYPSTGMLVLFFALHVCDEVNVYGFGADSRGNWHHYWENNRYAGEFRKTGVHDADFEAHIIDMLAKASKIEVYRGN

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service