Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

SAB1411175

Sigma-Aldrich

Anti-QARS antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

GLNRS, PRO2195

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 87.8 kDa

species reactivity

human, mouse, rat

technique(s)

immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... QARS(5859)

General description

Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. In metazoans, 9 aminoacyl-tRNA synthetases specific for glutamine (gln), glutamic acid (glu), and 7 other amino acids are associated within a multienzyme complex. Although present in eukaryotes, glutaminyl-tRNA synthetase (QARS) is absent from many prokaryotes, mitochondria, and chloroplasts, in which Gln-tRNA(Gln) is formed by transamidation of the misacylated Glu-tRNA(Gln). Glutaminyl-tRNA synthetase belongs to the class-I aminoacyl-tRNA synthetase family. (provided by RefSeq)

Immunogen

QARS (NP_005042.1, 1 a.a. ~ 775 a.a) full-length human protein.

Sequence
MAALDSLSLFTSLGLSEQKARETLKNSALSAQLREAATQAQQTLGSTIDKATGILLYGLASRLRDTRRLSFLVSYIASKKIHTEPQLSAALEYVRSHPLDPIDTVDFERECGVGVIVTPEQIEEAVEAAINRHRPQLLVERYHFNMGLLMGEARAVLKWADGKMIKNEVDMQVLHLLGPKLEADLEKKFKVAKARLEETDRRTAKDVVENGETADQTLSLMEQLRGEALKFHKPGENYKTPGYVVTPHTMNLLKQHLEITGGQVRTRFPPEPNGILHIGHAKAINFNFGYAKANNGICFLRFDDTNPEKEEAKFFTAICDMVAWLGYTPYKVTYASDYFDQLYAWAVELIRRGLAYVCHQRGEELKGHNTLPSPWRDRPMEESLLLFEAMRKGKFSEGEATLRMKLVMEDGKMDPVAYRVKYTPHHRTGDKWCIYPTYDYTHCLCDSIEHITHSLCTKEFQARRSSYFWLCNALDVYCPVQWEYGRLNLHYAVVSKRKILQLVATGAVRDWDDPRLFTLTALRRRGFPPEAINNFCARVGVTVAQTTMEPHLLEACVRDVLNDTAPRAMAVLESLRVIITNFPAAKSLDIQVPNFPADETKGFHQVPFAPIVFIERTDFKEEPEPGFKRLAWGQPVGLRHTGYVIELQHVVKGPSGCVESLEVTCRRADAGEKPKAFIHWVSQPLMCEVRLYERLFQHKNPEDPTEVPGGFLSDLNLASLHVVDAALVDCSVALAKPFDKFQFERLGYFSVDPDSHQGKLVFNRTVTLKEDPGKV

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service