Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1409454

Sigma-Aldrich

Anti-NANS antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

SAS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 40.3 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NANS(54187)

General description

This gene encodes an enzyme that functions in the biosynthetic pathways of sialic acids. In vitro, the encoded protein uses N-acetylmannosamine 6-phosphate and mannose 6-phosphate as substrates to generate phosphorylated forms of N-acetylneuraminic acid (Neu5Ac) and 2-keto-3-deoxy-D-glycero-D-galacto-nononic acid (KDN), respectively; however, it exhibits much higher activity toward the Neu5Ac phosphate product. In insect cells, expression of this gene results in Neu5Ac and KDN production. This gene is related to the E. coli sialic acid synthase gene neuB, and it can partially restore sialic acid synthase activity in an E. coli neuB-negative mutant. (provided by RefSeq)

Immunogen

NANS (NP_061819.2, 1 a.a. ~ 359 a.a) full-length human protein.

Sequence
MPLELELCPGRWVGGQHPCFIIAEIGQNHQGDLDVAKRMIRMAKECGADCAKFQKSELEFKFNRKALERPYTSKHSWGKTYGEHKRHLEFSHDQYRELQRYAEEVGIFFTASGMDEMAVEFLHELNVPFFKVGSGDTNNFPYLEKTAKKGRPMVISSGMQSMDTMKQVYQIVKPLNPNFCFLQCTSAYPLQPEDVNLRVISEYQKLFPDIPIGYSGHETGIAISVAAVALGAKVLERHITLDKTWKGSDHSASLEPGELAELVRSVRLVERALGSPTKQLLPCEMACNEKLGKSVVAKVKIPEGTILTMDMLTVKVGEPKGYPPEDIFNLVGKKVLVTVEEDDTIMEELVDNHGKKIKS

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service