Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1409327

Sigma-Aldrich

Monoclonal Anti-CITED1 antibody produced in mouse

clone 2H6, purified immunoglobulin, buffered aqueous solution

Synonym(s):

MSG1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2H6, monoclonal

form

buffered aqueous solution

mol wt

antigen 46.97 kDa

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CITED1(4435)

General description

This gene encodes a member of the CREB-binding protein/p300-interacting transactivator with Asp/Glu-rich C-terminal domain (CITED) family of proteins. The encoded protein, also known as melanocyte-specific gene 1, may function as a transcriptional coactivator and may play a role in pigmentation of melanocytes. Alternatively spliced transcript variants have been described. (provided by RefSeq)

Immunogen

CITED1 (AAH04240, 1 a.a. ~ 193 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MPTTSRPALDVKGGTSPAKEDANQEMSSVAYSNLAVKDRKAVAILHYPGVASNGTKASGAPTSSSGSPIGSPTTTPPTKPPSFNLHPAPHLLASMQLQKLNSQYQGMAAATPGQPGEAGPLQNWDFGAQAGGAESLSPSAGAQSPAIIDSDPVDEEVLMSLVVELGLDRANELPELWLGQNEFDFTADFPSSC

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service