Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1407290

Sigma-Aldrich

Anti-GTPBP5 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ10741, MGC29512, ObgH1, dJ1005F21.2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~44 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GTPBP5(26164)

General description

Small G proteins, such as GTPBP5, act as molecular switches that play crucial roles in the regulation of fundamental cellular processes such as protein synthesis, nuclear transport, membrane trafficking, and signal transduction (Hirano et al., 2006 [PubMed 17054726]).[supplied by OMIM

Immunogen

GTPBP5 (NP_056481, 1 a.a. ~ 406 a.a) full-length human protein.

Sequence
MAPARCFSARLRTVFQGVGHWALSTWAGLKPSRLLPQRASPRLLSVGRADLAKHQELPGKKLLSEKKLKRYFVDYRRVLVCGGNGGAGASCFHSEPRKEFGGPDGGDGGNGGHVILRVDQQVKSLSSVLSRYQGFSGEDGGSKNCFGRSGAVLYIRVPVGTLVKEGGRVVADLSCVGDEYIAALGGAGGKGNRFFLANNNRAPVTCTPGQPGQQRVLHLELKTVAHAGMVGFPNAGKSSLLRAISNARPAVASYPFTTLKPHVGIVHYEGHLQIAVADIPGIIRGAHQNRGLGSAFLRHIERCRFLLFVVDLSQPEPWTQVDDLKYELEMYEKGLSARPHAIVANKIDLPEAQANLSQLRDHLGQEVIVLSALTGENLEQLLLHLKVLYDAYAEAELGQGRQPLRW

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service