Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1406995

Sigma-Aldrich

Anti-CLEC10A antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CD301, CLECSF13, CLECSF14, HML, HML2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~35.4 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CLEC10A(10462)

General description

This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type 2 transmembrane protein may function as a cell surface antigen. Two transcript variants encoding distinct isoforms have been identified for this gene. (provided by RefSeq)

Immunogen

CLEC10A (NP_878910.1, 1 a.a. ~ 316 a.a) full-length human protein.

Sequence
MTRTYENFQYLENKVKVQGFKNGPLPLQSLLQRLCSGPCHLLLSLGLGLLLLVIICVVGFQNSKFQRDLVTLRTDFSNFTSNTVAEIQALTSQGSSLEETIASLKAEVEGFKQERQAGVSELQEHTTQKAHLGHCPHCPSVCVPVHSEMLLRVQQLVQDLKKLTCQVATLNNNASTEGTCCPVNWVEHQDSCYWFSHSGMSWAEAEKYCQLKNAHLVVINSREEQNFVQKYLGSAYTWMGLSDPEGAWKWVDGTDYATGFQNWKPGQPDDWQGHGLGGGEDCAHFHPDGRWNDDVCQRPYHWVCEAGLGQTSQESH

Application

Anti-CLEC10A antibody produced in mouse is suitable for western blot assay.

Biochem/physiol Actions

CLEC10A (C-type lectin domain family 10, member A) functions as a tumor associated macrophages in the tumor progression. It has been predicted that after being expressed by macrophages, glycoreceptor CLEC10A may directly interact with the glycan-binding receptors to identify the presence of certain glycan in human tumors. It has also been reported that macrophage cell surface lectin recognizes a common human carcinoma-associated epitope, Tn Ag.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Nollau et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 61(3), 199-205 (2013-01-01)
Specialized protein domains bind to posttranslational modifications (PTMs) of proteins, such as phosphorylation or glycosylation. When such PTM-binding protein domains are used as analytical tools, the functional states of cells and tissues can be determined with high precision. Here, we
N Suzuki et al.
Journal of immunology (Baltimore, Md. : 1950), 156(1), 128-135 (1996-01-01)
A human macrophage calcium-dependent (C-type) lectin cDNA clone was obtained from a library derived from IL-2-treated peripheral blood monocytes. The cDNA cloning was based on the structural homology to hepatic asialoglycoprotein receptors. The nucleotide sequence of this cDNA clone was
Toshiya Okumura et al.
PloS one, 9(6), e100559-e100559 (2014-06-20)
Phosphorylation of hormone-sensitive lipase (HSL) and perilipin by protein kinase A (PKA) promotes the hydrolysis of lipids in adipocytes. Although activation of lipolysis by PKA has been well studied, inactivation via protein phosphatases is poorly understood. Here, we investigated whether

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service