Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1404759

Sigma-Aldrich

Monoclonal Anti-ARHGEF11 antibody produced in mouse

clone 2H7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

DKFZp667F1223, GTRAP48, KIAA0380, PDZ-RHOGEF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2H7, monoclonal

form

buffered aqueous solution

mol wt

antigen ~37 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ARHGEF11(9826)

General description

Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli that work through G protein coupled receptors. The encoded protein may form a complex with G proteins and stimulate Rho-dependent signals. A similar protein in rat interacts with glutamate transporter EAAT4 and modulates its glutamate transport activity. Expression of the rat protein induces the reorganization of the actin cytoskeleton and its overexpression induces the formation of membrane ruffling and filopodia. Two alternative transcripts encoding different isoforms have been described. (provided by RefSeq)

Immunogen

ARHGEF11 (AAH57394, 651 a.a. ~ 750 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
RSESLKGREEMKRSRKAENVPRSRSDVDMDAAAEATRLHQSASSSTSSLSTRSLENPTPPFTPKMGRRSIESPSLGFCTDTLLPHLLEDDLGQLSDLEPE

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service