Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1404654

Sigma-Aldrich

Monoclonal Anti-RAB7L1 antibody produced in mouse

clone 1B10, purified immunoglobulin, buffered aqueous solution

Synonym(s):

DKFZp686P1051, RAB7L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1B10, monoclonal

form

buffered aqueous solution

mol wt

antigen ~48.44 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2bκ

NCBI accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAB7L1(8934)

General description

RAB7L1 (Rab-7-like protein 1, also referred to as RAB29) is a cytosolic GTPase. The gene encoding it is localized on human chromosome 1q32. Mouse monoclonal antibody raised against a full-length recombinant RAB7L1.

Immunogen

RAB7L1 (AAH02585, 1 a.a. ~ 203 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MGSRDHLFKVLVVGDAAVGKTSLVQRYSQDSFSKHYKSTVGVDFALKVLQWSDYEIVRLQLWDIAGQERFTSMTRLYYRDASACVIMFDVTNATTFSNSQRWKQDLDSKLTLPNGEPVPCLLLANKCDLSPWAVSRDQIDRFSKENGFTGWTETSVKENKNINEAMRVLIEKMMRNSTEDIMSLSTQGDYINLQTKSSSWSCC

Biochem/physiol Actions

RAB7L1 (Rab-7-like protein 1, also referred to as RAB29) modulates protein transport from the Golgi apparatus to lysosomes. It also modulates phagocytosis. RAB7L1 has been associated with Parkinson′s disease.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X-Y Guo et al.
European journal of neurology, 21(10), 1337-1343 (2014-07-22)
Recently, the rs1572931 single-nucleotide polymorphism (SNP) of the putative promoter of the member RAS oncogene family-like 1 (RAB7L1) gene was reported to be associated with reduced risk for Parkinson's disease (PD) in the Ashkenazi Jewish population. Ethnic-specific effects are an
Shicong Wang et al.
PloS one, 9(5), e96242-e96242 (2014-05-03)
Rab29 (also referred as Rab7L1) is a novel Rab protein, and is recently demonstrated to regulate phagocytosis and traffic from the Golgi to the lysosome. However, its roles in membrane trafficking have not been investigated extensively. Our results in this
David A MacLeod et al.
Neuron, 77(3), 425-439 (2013-02-12)
Recent genome-wide association studies have linked common variants in the human genome to Parkinson's disease (PD) risk. Here we show that the consequences of variants at 2 such loci, PARK16 and LRRK2, are highly interrelated, both in terms of their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service