Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1402987

Sigma-Aldrich

Monoclonal Anti-NIPSNAP1, (C-terminal) antibody produced in mouse

clone 3B7, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3B7, monoclonal

form

buffered aqueous solution

mol wt

antigen ~37.11 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NIPSNAP1(8508)

General description

NIPSNAP1 protein has a strong sequence similarity to the central portion of a hypothetical protein encoded by C. elegans chromosome III between a 4-nitrophenylphosphatase (NIP) domain and non-neuronal SNAP25-like protein. The NIPSNAP1 gene is located between the NF2 and pK1.3 genes on 22q12. (provided by RefSeq)

Immunogen

NIPSNAP1 (NP_003625.1, 185 a.a. ~ 284 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
YELRTYKLKPGTMIEWGNNWARAIKYRQENQEAVGGFFSQIGELYVVHHLWAYKDLQSREETRNAAWRKRGWDENVYYTVPLVRHMESRIMIPLKISPLQ

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service