Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

SAB1309053

[KO Validated]Anti-HADHA antibody produced in rabbit

new

[Knockout Validated],affinity isolated antibody, buffered aqueous solution

Synonym(s):

Trifunctional enzyme subunit alpha, mitochondrial, 78 kDa gastrin-binding protein, TP-alpha, Long-chain enoyl-CoA hydratase, Long chain 3-hydroxyacyl-CoA dehydrogenase, HADHA, HADH

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

(Human)

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

mol wt

27084Da

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:10~50
western blot: 1:1000
western blot: 1:500-1:2000

isotype

IgG

storage temp.

-10 to -25°C

target post-translational modification

unmodified

Gene Information

human ... HADHA(3030)

General description

HADHA is the alpha subunit of the mitochondrial trifunctional protein, which catalyzes the last three steps of mitochondrial beta-oxidation of long chain fatty acids. The mitochondrial membrane-bound heterocomplex is composed of four alpha and four beta subunits, with the alpha subunit catalyzing the 3-hydroxyacyl-CoA dehydrogenase and enoyl-CoA hydratase activities.

Specificity

This HADHA antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 737-763 amino acids from the C-terminal region of human HADHA.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide. This antibody is purified through a protein A column, followed by peptide affinity purification.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service