Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1305487

Sigma-Aldrich

MONOCLONAL ANTI-CD138 antibody produced in mouse

clone 480CT5.4.3, crude ascites, buffered aqueous solution

Synonym(s):

CD_antigen=CD138, Flags: Precursor, SDC, SDC1, Syndecan-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

crude ascites

antibody product type

primary antibodies

clone

480CT5.4.3, monoclonal

form

buffered aqueous solution

mol wt

32462 Da

species reactivity

human

technique(s)

western blot: 1:1000-1:8000

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SDC1(6382)

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiu-Feng Hu et al.
Molecular medicine reports, 10(3), 1409-1415 (2014-07-16)
The present study aimed to determine whether the expression levels of midkine (MK) and syndecan‑1 correlate with the malignant progression and poor prognosis of gastric cardiac adenocarcinoma (GCA). GCA tissue samples (n=72) were obtained from the Department of Pathology of
Laleh Talebian et al.
Transfusion, 54(6), 1515-1521 (2014-01-23)
The NKG2D receptor, one of the natural killer (NK) cell-activating receptors, is expressed on the surface of CD3+CD8+ T cells, γδ+ T cells, NK cells, NKT cells, and a few CD4+ T cells. We show, for the first time, a
Benjamin R Umiker et al.
European journal of immunology, 44(10), 3093-3108 (2014-07-22)
Systemic lupus erythematosus (SLE) is an autoimmune disease characterized by the presence of pathogenic IgG antinuclear antibodies. Pathogenic IgG autoantibody production requires B-cell activation, leading to the production of activation-induced deaminase (AID) and class switching of IgM genes to IgG.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service