Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300962

Sigma-Aldrich

Anti-Semaphorin 3E(Sema3e) (C-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Sema H, Anti-Sema3e, Anti-Semah, Anti-Semaphorin-3E, Anti-Semaphorin-H

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SEMA3E(9723)

General description

The Semaphorins make up the largest family of axon guidance cues yet described. Semaphorins are divided into 8 classes (classes 3-7 found in vertebrates). Class 3 Semaphorins are secreted, classes 4 through 6 are transmembrane proteins, and class 7 are membrane associated via glycosylphosphatidylinositol (GPI) linkage. They are characterized structurally by a conserved ~400 amino acid sema domain. They are classically described as collapsing factors and mediators of axon repulsion, although they may also act as context-dependent chemoattractants. Semaphorins have been shown to have roles in cardiovascular development and in the regulation of immune cell antigen presentation. Receptors or receptor complexes that mediate semaphorin signaling include proteins of the Neuropilin and Plexin families.

Immunogen

SEMAPHORIN 3E (NP_035478, 744-778)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-terminal region of mouse Sema3e.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service