S8686
SigmaScreen™ Streptavidin coated plates
384-well, clear
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352204
Recommended Products
Looking for similar products? Visit Product Comparison Guide
General description
SigmaScreen Streptavidin coated plates provide superior binding capacity with the greatest signal to noise ratios for improved sensitivity. With at least 6 pmol/well for the 96 well plates and approximately 4 pmol/well for the 384 well plates, SigmaScreen Streptavidin coated plates are pre-blocked with BSA and developed for maximum binding of biotinylated nucleic acids, antibodies, proteins and peptides. Manufactured under ISO9002 in Sigma′s GMP facility, Sigma′s Streptavidin coated plates deliver high well-to-well and plate-to-plate consistency.
Legal Information
SigmaScreen is a trademark of Sigma-Aldrich Co. LLC
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Pär Comstedt et al.
PloS one, 9(11), e113294-e113294 (2014-11-20)
There is currently no Lyme borreliosis vaccine available for humans, although it has been shown that the disease can be prevented by immunization with an OspA-based vaccine (LYMErix). Outer surface protein A (OspA) is one of the dominant antigens expressed
Articles
Review a high-throughput ELISA-based method to identify peptide substrates for class-specific and/or enzyme-specific Protein Tyrosine Kinases (PTKs) that can be utilized for the detection of kinase activity both in vivo and in vitro.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service