Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

S7944

Sigma-Aldrich

Anti-Sodium Channel ASIC1 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-ACCN2, Anti-Amiloride-sensitive Brain Sodium Channel, Anti-BnaC2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat, human, mouse

technique(s)

immunohistochemistry: suitable using rat brain secions
western blot: 1:200 using rat brain membranes

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACCN2(41)
mouse ... Accn2(11419)
rat ... Accn2(79123)

Specificity

Recognizes isoforms ASIC1α and ASIC1β.

Immunogen

synthetic peptide corresponding to residues 469-488 of rat ASIC1.

Target description

Sodium Channel ASIC1 encodes a member of the degenerin/epithelial sodium channel (DEG/ENaC) superfamily. The members of this family are amiloride-sensitive sodium channels that contain intracellular N and C termini, 2 hydrophobic transmembrane regions, an

Physical form

Lyophilized from phosphate buffered saline (PBS), pH 7.4, 1% BSA, 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephan Kellenberger et al.
Physiological reviews, 82(3), 735-767 (2002-06-28)
The recently discovered epithelial sodium channel (ENaC)/degenerin (DEG) gene family encodes sodium channels involved in various cell functions in metazoans. Subfamilies found in invertebrates or mammals are functionally distinct. The degenerins in Caenorhabditis elegans participate in mechanotransduction in neuronal cells
Oleg Krishtal
Trends in neurosciences, 26(9), 477-483 (2003-09-02)
Acid-sensing ionic channels (ASICs) are almost ubiquitous in the mammalian nervous system, both at the periphery and in the brain. Strong evidence for the physiological function of these molecules has come from recent knockout experiments. Now it is clear that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service